siRNA Details
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 4, 5 | 530 | ||
NR_073103.1 | 4, 5 | 530 | ||||
NR_073104.1 | 4, 5 | 530 | ||||
NR_073105.1 | 4, 5 | 530 | ||||
NR_073106.1 | 4, 5 | 530 | ||||
NR_073107.1 | 3, 4 | 409 | ||||
GenBank mRNA | AK307952.1 | - | 4, 5 | 519 | ||
BC143505.1 | - | 4, 5 | 416 | |||
BC143508.1 | - | 4, 5 | 416 | |||
BC143509.1 | - | 4, 5 | 416 | |||
BC143511.1 | - | 4, 5 | 416 | |||
BC143512.1 | - | 3, 4 | 295 | |||
BP280760.1 | - | 4, 5 | 533 | |||
X17033.1 | - | 4, 5 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
