siRNA Details
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 30 | 3424 | ||
GenBank mRNA | BC008786.2 | - | 30 | 3409 | 3629647 | |
BX647780.1 | - | 17 | 4170 | |||
M13918.2 | - | 30 | 3311 | |||
X06256.1 | - | 30 | 3379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
