siRNA Details
Gene Symbol: ITPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002223.3 | NP_002214.2 | 5, 6 | 941 | ||
XM_017019266.1 | XP_016874755.1 | 5, 6 | 940 | |||
XM_017019267.1 | XP_016874756.1 | 5, 6 | 464 | |||
XM_017019269.1 | XP_016874758.1 | 5, 6 | 940 | |||
GenBank mRNA | AK307712.1 | - | 5, 6 | 817 | ||
AK310504.1 | - | 5, 6 | 681 | |||
AY429550.1 | - | 4, 5 | 458 | |||
BX495410.1 | - | 1, 2 | 161 | |||
D26350.1 | - | 5, 6 | 942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
