siRNA Details
Gene Symbol: ITPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002224.3 | NP_002215.2 | 3 | 451 | ||
XM_011514577.2 | XP_011512879.1 | 2 | 91 | |||
XM_017010832.1 | XP_016866321.1 | 4 | 280 | |||
GenBank mRNA | BC146646.1 | - | 3 | 410 | ||
D26351.1 | - | 3 | 462 | |||
U01062.1 | - | 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
