siRNA Details
Gene Symbol: MEST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253901.1 | NP_001240830.1 | 2 | 362 | ||
NM_001253902.1 | NP_001240831.1 | 2 | 314 | |||
NM_002402.3 | NP_002393.2 | 2 | 423 | |||
NM_177524.2 | NP_803490.1 | 2 | 362 | |||
NM_177525.2 | NP_803491.1 | 2 | 299 | |||
XM_011516222.1 | XP_011514524.1 | 2 | 409 | |||
XM_017012218.1 | XP_016867707.1 | 2 | 202 | |||
GenBank mRNA | AK098397.1 | - | 2 | 332 | ||
AK312688.1 | - | 2 | 338 | |||
BC002413.2 | - | 2 | 384 | 2964732 | ||
BC011908.2 | - | 2 | 388 | 4137253 | ||
BC014564.2 | - | 2 | 384 | 2964732 | ||
BC018695.2 | - | 2 | 384 | 2964732 | ||
BC090049.1 | - | 2 | 183 | 30379091 | ||
BG831799.1 | - | 2 | 363 | |||
BI459412.1 | - | 2 | 296 | |||
BT007056.1 | - | 2 | 167 | |||
CR457040.1 | - | 2 | 167 | |||
CR627364.1 | - | 2 | 250 | |||
D78611.1 | - | 2 | 390 | |||
D87367.1 | - | 2 | 389 | |||
DA487705.1 | - | 2 | 314 | |||
DB454206.1 | - | 2 | 301 | |||
Y11534.1 | - | 2 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
