siRNA Details
Gene Symbol: MEST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253900.1 | NP_001240829.1 | 12 | 1408 | ||
NM_001253901.1 | NP_001240830.1 | 11 | 1287 | |||
NM_001253902.1 | NP_001240831.1 | 11 | 1239 | |||
NM_002402.3 | NP_002393.2 | 12 | 1450 | |||
NM_177524.2 | NP_803490.1 | 12 | 1389 | |||
NM_177525.2 | NP_803491.1 | 12 | 1326 | |||
XM_011516222.1 | XP_011514524.1 | 11 | 1334 | |||
XM_017012218.1 | XP_016867707.1 | 12 | 1229 | |||
GenBank mRNA | AK098397.1 | - | 12 | 1359 | ||
BC002413.2 | - | 12 | 1411 | 2964732 | ||
BC011908.2 | - | 12 | 1415 | 4137253 | ||
BC014564.2 | - | 12 | 1411 | 2964732 | ||
BC018695.2 | - | 12 | 1411 | 2964732 | ||
BC090049.1 | - | 11 | 1108 | 30379091 | ||
CR627364.1 | - | 12 | 1277 | |||
D78611.1 | - | 12 | 1417 | |||
D87367.1 | - | 12 | 1417 | |||
Y11534.1 | - | 12 | 1412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
