siRNA Details
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002507.3 | NP_002498.1 | 6 | 3031 | ||
GenBank mRNA | BC050309.1 | - | 6 | 3031 | 5263715 | |
BQ639556.1 | - | 1 | 419 | |||
M14764.1 | - | 6 | 3019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
