siRNA Details
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002546.3 | NP_002537.3 | 4 | 954 | ||
GenBank mRNA | AB002146.1 | - | 4 | 631 | ||
AF134187.1 | - | 3 | 569 | |||
AK223155.1 | - | 4 | 693 | |||
AK308524.1 | - | 5 | 975 | |||
AK313710.1 | - | 4 | 1048 | |||
BC030155.2 | - | 4 | 852 | 4875170 | ||
U94332.1 | - | 4 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
