siRNA Details
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002644.3 | NP_002635.2 | 3 | 434 | ||
XM_011509629.1 | XP_011507931.1 | 3 | 345 | |||
GenBank mRNA | AF272149.2 | - | 3 | 430 | ||
AK225471.1 | - | 3 | 434 | |||
BC110494.2 | - | 3 | 331 | 40007650 | ||
BC110495.2 | - | 3 | 331 | 40007652 | ||
CR749533.1 | - | 4 | 507 | |||
M24559.1 | - | 1 | 37 | |||
S62403.1 | - | 3 | 406 | |||
X73079.1 | - | 3 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
