siRNA Details
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002644.3 | NP_002635.2 | 11 | 3168 | ||
XM_011509629.1 | XP_011507931.1 | 11 | 3079 | |||
GenBank mRNA | AF272149.2 | - | 11 | 3164 | ||
AK310199.1 | - | 1 | 617 | |||
AY178835.1 | - | 1 | 692 | |||
CR749533.1 | - | 12 | 3241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
