siRNA Details
Gene Symbol: NECTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042724.1 | NP_001036189.1 | 2 | 494 | ||
NM_002856.2 | NP_002847.1 | 2 | 494 | |||
XM_011527192.2 | XP_011525494.1 | 2 | 494 | |||
GenBank mRNA | AF058448.1 | - | 2 | 253 | ||
AK291330.1 | - | 2 | 320 | |||
BC003091.1 | - | 2 | 302 | 3503222 | ||
CN410857.1 | - | 2 | 466 | |||
CR456818.1 | - | 2 | 104 | |||
X80038.1 | - | 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
