siRNA Details
Gene Symbol: NECTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002856.2 | NP_002847.1 | 5, 6 | 1424 | ||
GenBank mRNA | AF058448.1 | - | 5, 6 | 1183 | ||
AK291330.1 | - | 5, 6 | 1250 | |||
BC003091.1 | - | 5, 6 | 1232 | 3503222 | ||
CR456818.1 | - | 5, 6 | 1034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
