siRNA Details
Gene Symbol: ROBO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145845.1 | NP_001139317.1 | 2 | 1078 | ||
NM_002941.3 | NP_002932.1 | 4 | 405 | |||
NM_133631.3 | NP_598334.2 | 2 | 1078 | |||
XM_006713277.2 | XP_006713340.1 | 2 | 1047 | |||
XM_011533976.1 | XP_011532278.1 | 4 | 494 | |||
XM_011533977.2 | XP_011532279.1 | 4 | 347 | |||
XM_011533978.1 | XP_011532280.1 | 4 | 494 | |||
XM_011533979.1 | XP_011532281.1 | 4 | 494 | |||
XM_011533980.1 | XP_011532282.1 | 4 | 494 | |||
XM_017006982.1 | XP_016862471.1 | 3 | 406 | |||
XM_017006983.1 | XP_016862472.1 | 2 | 1047 | |||
XM_017006984.1 | XP_016862473.1 | 4 | 494 | |||
XM_017006985.1 | XP_016862474.1 | 2 | 147 | |||
GenBank mRNA | AF040990.1 | - | 3 | 197 | ||
AK297481.1 | - | 3 | 353 | |||
BC112336.1 | - | 2 | 131 | 40037251 | ||
BC115020.1 | - | 2 | 179 | 7961997 | ||
BC115022.1 | - | 2 | 179 | 7962000 | ||
BC157861.1 | - | 2 | 375 | 9057080 | ||
BC171855.1 | - | 2 | 375 | 9054509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
