siRNA Details
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 1, 2 | 410 | ||
XM_011542963.2 | XP_011541265.1 | 1, 2 | 285 | |||
XM_017018171.1 | XP_016873660.1 | 1, 2 | 285 | |||
GenBank mRNA | AK309523.1 | - | 1, 2 | 233 | ||
BC137171.1 | - | 1, 2 | 333 | 9021168 | ||
BX365721.2 | - | 1, 2 | 411 | |||
Y08110.1 | - | 1, 2 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
