siRNA Details
Gene Symbol: SRPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177842.1 | NP_001171313.1 | 3, 4 | 624 | ||
NM_003139.3 | NP_003130.2 | 4, 5 | 708 | |||
XM_017018179.1 | XP_016873668.1 | 4, 5 | 654 | |||
GenBank mRNA | AK130382.1 | - | 1, 2 | 123 | ||
AK303379.1 | - | 3, 4 | 624 | |||
AK312377.1 | - | 4, 5 | 626 | |||
BC001162.1 | - | 4, 5 | 588 | 3528505 | ||
BC008077.2 | - | 4, 5 | 587 | |||
BC009110.1 | - | 4, 5 | 551 | 3453825 | ||
BC013583.1 | - | 4, 5 | 614 | 3877676 | ||
CR457025.1 | - | 4, 5 | 530 | |||
X06272.1 | - | 4, 5 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
