siRNA Details
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 4, 5 | 580 | ||
NM_003227.3 | NP_003218.2 | 7, 8 | 1000 | |||
XM_005250553.4 | XP_005250610.1 | 8, 9 | 1306 | |||
XM_017012573.1 | XP_016868062.1 | 8, 9 | 1108 | |||
GenBank mRNA | AF067864.1 | - | 7, 8 | 986 | ||
AF085928.1 | - | 4, 5 | 443 | |||
AK000421.1 | - | 2, 3 | 214 | |||
BC142630.1 | - | 7, 8 | 986 | 40147690 | ||
BX384750.2 | - | 4, 5 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
