siRNA Details
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 3 | 332 | ||
NM_003234.3 | NP_003225.2 | 3 | 473 | |||
GenBank mRNA | AB209254.1 | - | 3 | 236 | ||
AK291723.1 | - | 3 | 473 | |||
AK309923.1 | - | 3 | 261 | |||
BC001188.1 | - | 3 | 246 | 3354176 | ||
CF994006.1 | - | 3 | 360 | |||
DA460386.1 | - | 3 | 332 | |||
M11507.1 | - | 3 | 285 | |||
X01060.1 | - | 3 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
