siRNA Details
Gene Symbol: TGFBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195683.1 | NP_001182612.1 | 7 | 1270 | ||
NM_001195684.1 | NP_001182613.1 | 8 | 1106 | |||
NM_003243.4 | NP_003234.2 | 7 | 1270 | |||
NR_036634.1 | 8 | 1367 | ||||
XM_006710867.2 | XP_006710930.1 | 7 | 989 | |||
XM_006710868.2 | XP_006710931.1 | 7 | 955 | |||
GenBank mRNA | AJ251961.1 | - | 6 | 817 | ||
AK225799.1 | - | 7 | 1048 | |||
AK291345.1 | - | 7 | 911 | |||
BC099914.1 | - | 10 | 1509 | |||
BC126116.1 | - | 7 | 923 | 8991832 | ||
BC136295.1 | - | 7 | 923 | 9020282 | ||
BC143869.1 | - | 8 | 1020 | |||
HG764816.1 | - | 8 | 1106 | |||
L07594.1 | - | 8 | 1100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
