siRNA Details
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318787.1 | NP_001305716.1 | 4 | 634 | ||
NM_001318789.1 | NP_001305718.1 | 3 | 424 | |||
NM_001318790.1 | NP_001305719.1 | 3 | 401 | |||
NM_001318791.1 | NP_001305720.1 | 3 | 390 | |||
NM_001318793.1 | NP_001305722.1 | 3 | 386 | |||
NM_001318795.1 | NP_001305724.1 | 3 | 365 | |||
NM_001318796.1 | NP_001305725.1 | 3 | 350 | |||
NM_003264.4 | NP_003255.2 | 3 | 409 | |||
XM_011532215.2 | XP_011530517.1 | 3 | 334 | |||
XM_011532216.2 | XP_011530518.1 | 2 | 281 | |||
XM_017008573.1 | XP_016864062.1 | 4 | 880 | |||
XM_017008574.1 | XP_016864063.1 | 4 | 562 | |||
XM_017008575.1 | XP_016864064.1 | 4 | 606 | |||
XM_017008576.1 | XP_016864065.1 | 2 | 1180 | |||
GenBank mRNA | AB445624.1 | - | 1 | 153 | ||
AF051152.1 | - | 2 | 315 | |||
AF502291.1 | - | 1 | 153 | |||
AK125659.1 | - | 1 | 1557 | |||
BC033756.1 | - | 3 | 303 | 5213439 | ||
BI769109.1 | - | 3 | 372 | |||
DA460298.1 | - | 4 | 375 | |||
DA870971.1 | - | 4 | 390 | |||
DQ012265.1 | - | 1 | 153 | |||
DQ012266.1 | - | 1 | 153 | |||
DQ012267.1 | - | 1 | 153 | |||
DQ012268.1 | - | 1 | 153 | |||
DQ012269.1 | - | 1 | 153 | |||
DQ012270.1 | - | 1 | 153 | |||
DQ012271.1 | - | 1 | 153 | |||
KU178364.1 | - | 1 | 153 | |||
KU178365.1 | - | 1 | 153 | |||
U88878.1 | - | 2 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
