siRNA Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003265.2 | NP_003256.1 | 2 | 327 | ||
GenBank mRNA | AB445631.1 | - | 1 | 226 | ||
AK314208.1 | - | 2 | 326 | |||
BC059372.1 | - | 2 | 326 | |||
BC094737.1 | - | 2 | 326 | 30531515 | ||
BC096333.1 | - | 2 | 304 | 40008349 | ||
BC096334.3 | - | 2 | 304 | 40008350 | ||
BC096335.1 | - | 2 | 304 | 40008351 | ||
DA374150.1 | - | 2 | 327 | |||
DQ445682.1 | - | 2 | 297 | |||
U88879.1 | - | 2 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
