siRNA Details
Gene Symbol: TNFRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003327.3 | NP_003318.1 | 3 | 402 | ||
XM_011542074.2 | XP_011540376.1 | 1 | 756 | |||
XM_011542075.2 | XP_011540377.1 | 3 | 869 | |||
XM_011542076.2 | XP_011540378.1 | 1 | 756 | |||
XM_017002231.1 | XP_016857720.1 | 4 | 1747 | |||
XM_017002232.1 | XP_016857721.1 | 4 | 1747 | |||
GenBank mRNA | BC105070.1 | - | 3 | 402 | 8144073 | |
BC105072.1 | - | 3 | 402 | 8144075 | ||
S76792.1 | - | 3 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
