siRNA Details
Gene Symbol: TBCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033115.4 | NP_149106.2 | 3, 4 | 627 | ||
XM_017008849.1 | XP_016864338.1 | 3, 4 | 303 | |||
GenBank mRNA | BC009208.2 | - | 3, 4 | 589 | 3634845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
