siRNA Details
Gene Symbol: PPFIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003660.3 | NP_003651.1 | 3 | 618 | ||
NR_103842.1 | 3 | 618 | ||||
XM_017027407.1 | XP_016882896.1 | 3 | 618 | |||
GenBank mRNA | AB014554.2 | - | 3 | 601 | ||
AK289757.1 | - | 3 | 323 | |||
AK293798.1 | - | 1 | 69 | |||
AK308483.1 | - | 1 | 109 | |||
BC101518.1 | - | 3 | 344 | 8069024 | ||
BC101520.1 | - | 3 | 344 | 8069026 | ||
BC143470.1 | - | 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
