siRNA Details
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 3 | 604 | ||
NM_147187.2 | NP_671716.2 | 3 | 604 | |||
NR_027140.1 | 2 | 498 | ||||
XM_017013944.1 | XP_016869433.1 | 3 | 392 | |||
GenBank mRNA | AB451272.1 | - | 3 | 311 | ||
AB451399.1 | - | 3 | 311 | |||
AF012535.1 | - | 3 | 450 | |||
AF012628.1 | - | 3 | 311 | |||
AF016266.1 | - | 3 | 428 | |||
AF016268.1 | - | 3 | 420 | |||
AF016849.1 | - | 3 | 455 | |||
AF018657.1 | - | 3 | 311 | |||
AF018658.1 | - | 3 | 311 | |||
AF020501.1 | - | 3 | 311 | |||
AF022386.1 | - | 3 | 311 | |||
AF153687.1 | - | 3 | 429 | |||
AF192548.1 | - | 3 | 411 | |||
AK291216.1 | - | 3 | 431 | |||
AK298602.1 | - | 2 | 342 | |||
AL529460.3 | - | 3 | 604 | |||
AY358277.1 | - | 3 | 450 | |||
BC001281.1 | - | 3 | 458 | 3458466 | ||
BX538104.1 | - | 3 | 444 | |||
CR541898.1 | - | 3 | 311 | |||
DA738472.1 | - | 3 | 604 | |||
DA889488.1 | - | 3 | 448 | |||
FJ200482.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
