siRNA Details
Gene Symbol: P4HA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142599.1 | NP_001136071.1 | 13 | 1550 | ||
NM_004199.2 | NP_004190.1 | 12 | 1886 | |||
XM_005272116.4 | XP_005272173.1 | 12 | 1414 | |||
XM_005272117.4 | XP_005272174.1 | 13 | 1572 | |||
XM_005272118.4 | XP_005272175.1 | 13 | 1701 | |||
XM_005272119.4 | XP_005272176.1 | 13 | 1891 | |||
XM_005272120.4 | XP_005272177.1 | 13 | 1671 | |||
XM_006714728.3 | XP_006714791.1 | 13 | 1948 | |||
XM_011543705.2 | XP_011542007.1 | 10 | 1163 | |||
GenBank mRNA | U90441.1 | - | 12 | 1509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
