siRNA Details
Gene Symbol: IL1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261419.1 | NP_001248348.1 | 3 | 335 | ||
NM_004633.3 | NP_004624.1 | 3 | 335 | |||
NR_048564.1 | 3 | 322 | ||||
XM_006712734.3 | XP_006712797.1 | 3 | 359 | |||
XM_006712736.3 | XP_006712799.1 | 3 | 396 | |||
XM_011511801.2 | XP_011510103.1 | 3 | 2695 | |||
XM_011511804.2 | XP_011510106.1 | 3 | 388 | |||
XM_011511807.1 | XP_011510109.1 | 3 | 2695 | |||
XM_017004889.1 | XP_016860378.1 | 2 | 245 | |||
GenBank mRNA | AK130412.1 | - | 3 | 219 | ||
AK309161.1 | - | 2 | 91 | |||
BC027733.2 | - | 3 | 289 | |||
BC039031.1 | - | 3 | 335 | 5723005 | ||
U64094.1 | - | 2 | 167 | |||
U74649.1 | - | 2 | 141 | |||
X59770.1 | - | 2 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
