siRNA Details
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004871.2 | NP_004862.1 | 2, 3 | 180 | ||
XM_005258070.4 | XP_005258127.1 | 2, 3 | 421 | |||
XM_005258072.2 | XP_005258129.1 | 2, 3 | 208 | |||
XM_017025374.1 | XP_016880863.1 | 2, 3 | 424 | |||
XM_017025375.1 | XP_016880864.1 | 2, 3 | 183 | |||
XM_017025377.1 | XP_016880866.1 | 2, 3 | 430 | |||
GenBank mRNA | AF073926.1 | - | 2, 3 | 166 | ||
AK000606.1 | - | 2, 3 | 180 | |||
AK291381.1 | - | 2, 3 | 180 | |||
BC012620.1 | - | 2, 3 | 169 | 4250494 | ||
BC040471.1 | - | 2, 3 | 161 | 4697067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
