siRNA Details
Gene Symbol: ITGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002204.3 | NP_002195.1 | 2 | 780 | ||
XM_005257308.2 | XP_005257365.1 | 2 | 683 | |||
GenBank mRNA | AB209658.1 | - | 2 | 680 | ||
AK095140.1 | - | 1 | 641 | |||
AK289961.1 | - | 2 | 406 | |||
AK293323.1 | - | 3 | 360 | |||
AK303384.1 | - | 3 | 334 | |||
BC136636.1 | - | 2 | 380 | 9020626 | ||
BC144328.1 | - | 2 | 380 | 9052848 | ||
BC150190.1 | - | 2 | 623 | 40148521 | ||
D01038.1 | - | 2 | 212 | |||
M59911.1 | - | 2 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
