siRNA Details
Gene Symbol: SCARB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082959.1 | NP_001076428.1 | 4 | 867 | ||
NM_005505.4 | NP_005496.4 | 4 | 867 | |||
GenBank mRNA | AB209436.1 | - | 4 | 698 | ||
AF515445.1 | - | 5 | 929 | |||
AK124112.1 | - | 4 | 916 | |||
AK302779.1 | - | 4 | 559 | |||
AK304730.1 | - | 4 | 757 | |||
BC080647.1 | - | 4 | 741 | 6576008 | ||
BC093732.1 | - | 4 | 677 | 7939577 | ||
BC112037.1 | - | 4 | 689 | 8327505 | ||
BC143319.1 | - | 4 | 677 | 9051824 | ||
KU177964.1 | - | 4 | 614 | |||
KU177965.1 | - | 4 | 614 | |||
Z22555.1 | - | 4 | 683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
