siRNA Details
Gene Symbol: SCARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204255.1 | NP_001191184.1 | 2 | 495 | ||
NM_005506.3 | NP_005497.1 | 2 | 495 | |||
GenBank mRNA | AK296519.1 | - | 2 | 199 | ||
AK313016.1 | - | 2 | 205 | |||
BC021892.1 | - | 2 | 161 | 3872778 | ||
BQ924546.1 | - | 2 | 478 | |||
BT006939.1 | - | 2 | 139 | |||
D12676.1 | - | 2 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
