siRNA Details
Gene Symbol: SCARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204255.1 | NP_001191184.1 | 5 | 998 | ||
NM_005506.3 | NP_005497.1 | 8 | 1427 | |||
GenBank mRNA | AK296519.1 | - | 5 | 702 | ||
AK313016.1 | - | 8 | 1137 | |||
BC021892.1 | - | 8 | 1093 | 3872778 | ||
BT006939.1 | - | 8 | 1071 | |||
D12676.1 | - | 8 | 1322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
