siRNA Details
Gene Symbol: IFNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005534.3 | NP_005525.2 | 7 | 1932 | ||
XM_005260969.2 | XP_005261026.1 | 8 | 1448 | |||
XM_011529553.1 | XP_011527855.1 | 8 | 1374 | |||
XM_011529554.2 | XP_011527856.1 | 7 | 1808 | |||
GenBank mRNA | BC003624.2 | - | 7 | 1399 | 2967074 | |
U05875.1 | - | 7 | 1933 | |||
U05877.1 | - | 7 | 1397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
