siRNA Details
Gene Symbol: KLRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005810.3 | NP_005801.3 | 4 | 403 | ||
XM_017018682.1 | XP_016874171.1 | 4 | 948 | |||
XM_017018683.1 | XP_016874172.1 | 4 | 948 | |||
XM_017018684.1 | XP_016874173.1 | 4 | 948 | |||
XM_017018685.1 | XP_016874174.1 | 4 | 948 | |||
GenBank mRNA | AF034952.1 | - | 4 | 392 | ||
AF081675.1 | - | 4 | 426 | |||
AF097358.1 | - | 4 | 409 | |||
AK301367.1 | - | 4 | 405 | |||
AK316337.1 | - | 4 | 487 | |||
BC012621.1 | - | 4 | 411 | 4108843 | ||
BQ212545.1 | - | 2 | 147 | |||
DT217650.1 | - | 4 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
