siRNA Details
Gene Symbol: RAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005856.2 | NP_005847.1 | 3 | 362 | ||
XM_006715631.3 | XP_006715694.1 | 5 | 685 | |||
XM_017011666.1 | XP_016867155.1 | 3 | 726 | |||
GenBank mRNA | AJ001016.1 | - | 3 | 330 | ||
AK128765.1 | - | 1 | 1692 | |||
AK311871.1 | - | 3 | 301 | |||
AY265459.1 | - | 3 | 301 | |||
BC022304.1 | - | 3 | 329 | 4717934 | ||
BC053852.1 | - | 3 | 328 | 6197363 | ||
BP347072.1 | - | 3 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
