siRNA Details
Gene Symbol: SIGMAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282205.1 | NP_001269134.1 | 4 | 722 | ||
NM_001282206.1 | NP_001269135.1 | 4 | 893 | |||
NM_001282207.1 | NP_001269136.1 | 4 | 880 | |||
NM_001282208.1 | NP_001269137.1 | 4 | 963 | |||
NM_001282209.1 | NP_001269138.1 | 3 | 800 | |||
NM_005866.3 | NP_005857.1 | 4 | 940 | |||
NM_147157.2 | NP_671513.1 | 3 | 847 | |||
NR_104108.1 | 4 | 827 | ||||
GenBank mRNA | AK098451.1 | - | 5 | 731 | ||
AK130502.1 | - | 4 | 923 | |||
AK222899.1 | - | 4 | 869 | |||
AK291092.1 | - | 3 | 776 | |||
AK299130.1 | - | 4 | 649 | |||
BC004899.2 | - | 4 | 846 | 3529352 | ||
BC007839.2 | - | 4 | 810 | |||
U75283.1 | - | 4 | 858 | |||
U79528.1 | - | 4 | 884 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
