siRNA Details
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005894.2 | NP_005885.1 | 3 | 415 | ||
XM_005245602.1 | XP_005245659.1 | 3 | 415 | |||
XM_017002806.1 | XP_016858295.1 | 3 | 415 | |||
GenBank mRNA | AF011429.1 | - | 3 | 335 | ||
AK292040.1 | - | 3 | 336 | |||
AY358494.1 | - | 3 | 369 | |||
BC033586.1 | - | 3 | 303 | 5225975 | ||
DA957550.1 | - | 3 | 415 | |||
U82812.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
