siRNA Details
Gene Symbol: PDGFRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006207.2 | NP_006198.1 | 5 | 964 | ||
GenBank mRNA | AK289450.1 | - | 5 | 964 | ||
BC010927.1 | - | 4 | 535 | 4278882 | ||
CR541952.1 | - | 4 | 519 | |||
D37965.1 | - | 4 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
