siRNA Details
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006404.4 | NP_006395.2 | 3 | 775 | ||
XM_011528496.1 | XP_011526798.1 | 3 | 618 | |||
GenBank mRNA | AK314887.1 | - | 3 | 741 | ||
BC014451.1 | - | 3 | 639 | 4907433 | ||
BM808358.1 | - | 3 | 793 | |||
BQ939781.1 | - | 1 | 107 | |||
CR456948.1 | - | 3 | 557 | |||
L35545.1 | - | 3 | 581 | |||
X89079.1 | - | 3 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
