siRNA Details
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135768.2 | NP_001129240.1 | 8 | 1569 | ||
NM_001135769.2 | NP_001129241.1 | 7 | 1545 | |||
NM_006505.4 | NP_006496.4 | 8 | 1704 | |||
GenBank mRNA | AK094177.1 | - | 8 | 1511 | ||
AK290946.1 | - | 8 | 1618 | |||
AK300349.1 | - | 8 | 1457 | |||
BC015542.1 | - | 8 | 1576 | 3902226 | ||
DA849917.1 | - | 3 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
