siRNA Details
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002167.4 | NP_002158.3 | 3 | 910 | ||
GenBank mRNA | A17546.1 | - | 3 | 665 | ||
AK225965.1 | - | 1 | 1271 | |||
BC003107.1 | - | 3 | 628 | 3543936 | ||
X66924.1 | - | 3 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
