siRNA Details
Gene Symbol: KDELR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006801.2 | NP_006792.1 | 2, 3 | 395 | ||
GenBank mRNA | AB097047.1 | - | 2, 3 | 393 | ||
AK075185.1 | - | 1, 2 | 440 | |||
AK312647.1 | - | 2, 3 | 407 | |||
BC008958.2 | - | 2, 3 | 383 | 3009767 | ||
BC018778.1 | - | 2, 3 | 395 | 4870476 | ||
X55885.1 | - | 2, 3 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
