siRNA Details
Gene Symbol: LILRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081442.2 | NP_001074911.1 | 3, 4 | 478 | ||
NM_006840.4 | NP_006831.1 | 3, 4 | 478 | |||
XM_011526359.2 | XP_011524661.1 | 3, 4 | 462 | |||
XM_011526360.2 | XP_011524662.1 | 3, 4 | 462 | |||
XM_011526361.2 | XP_011524663.1 | 3, 4 | 462 | |||
GenBank mRNA | AF025534.1 | - | 3, 4 | 429 | ||
AK223296.1 | - | 3, 4 | 434 | |||
AK290845.1 | - | 3, 4 | 462 | |||
BC025704.1 | - | 3, 4 | 432 | 5223581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
