siRNA Details
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100603.1 | NP_001094073.1 | 4 | 621 | ||
NM_006854.3 | NP_006845.1 | 5 | 874 | |||
GenBank mRNA | AK093917.1 | - | 3 | 1173 | ||
AL547016.3 | - | 5 | 851 | |||
BC008081.2 | - | 5 | 778 | 2966703 | ||
BC012994.2 | - | 5 | 809 | 3604062 | ||
BC071982.1 | - | 4 | 575 | 5477638 | ||
M88458.1 | - | 5 | 752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
