siRNA Details
Gene Symbol: HNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006895.2 | NP_008826.1 | 6 | 1363 | ||
XM_011511064.2 | XP_011509366.1 | 5 | 1126 | |||
XM_017003948.1 | XP_016859437.1 | 7 | 1823 | |||
GenBank mRNA | AF523359.1 | - | 6 | 1290 | ||
AF523360.1 | - | 6 | 1290 | |||
BC020677.1 | - | 6 | 1122 | 4767331 | ||
D16224.1 | - | 6 | 1119 | |||
U08092.1 | - | 6 | 1148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
