siRNA Details
Gene Symbol: XPOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007235.4 | NP_009166.2 | 3, 4 | 648 | ||
XM_017018748.1 | XP_016874237.1 | 3, 4 | 508 | |||
GenBank mRNA | AF039022.1 | - | 2, 3 | 142 | ||
AK292884.1 | - | 3, 4 | 461 | |||
AK308108.1 | - | 3, 4 | 469 | |||
BC020569.1 | - | 2, 3 | 227 | 4420052 | ||
BQ215290.1 | - | 3, 4 | 568 | |||
CR979468.1 | - | 3, 4 | 648 | |||
Y16414.1 | - | 2, 3 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
