siRNA Details
Gene Symbol: CD93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012072.3 | NP_036204.2 | 1 | 519 | ||
GenBank mRNA | AB209895.1 | - | 1 | 498 | ||
AJ295142.1 | - | 1 | 493 | |||
AK299957.1 | - | 1 | 503 | |||
BC028075.1 | - | 1 | 498 | 5216061 | ||
U94333.1 | - | 1 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
