siRNA Details
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173393.2 | NP_001166864.1 | 3 | 438 | ||
NM_001308156.1 | NP_001295085.1 | 3 | 559 | |||
NM_012206.3 | NP_036338.2 | 2 | 525 | |||
XM_017009339.1 | XP_016864828.1 | 2 | 239 | |||
XM_017009340.1 | XP_016864829.1 | 2 | 239 | |||
GenBank mRNA | AF043724.1 | - | 3 | 278 | ||
AK298259.1 | - | 3 | 559 | |||
BC013325.1 | - | 2 | 395 | 3826378 | ||
CR457114.1 | - | 2 | 227 | |||
DA384612.1 | - | 3 | 282 | |||
HQ412639.1 | - | 2 | 227 | |||
HQ412640.1 | - | 2 | 227 | |||
HQ412641.1 | - | 2 | 227 | |||
JX049978.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
