siRNA Details
Gene Symbol: NPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161363.1 | NP_001154835.1 | 4 | 838 | ||
NM_001161364.1 | NP_001154836.1 | 3 | 490 | |||
NM_012428.3 | NP_036560.1 | 4 | 838 | |||
NM_017455.3 | NP_059429.1 | 3 | 490 | |||
XM_011521452.2 | XP_011519754.1 | 4 | 703 | |||
GenBank mRNA | AB209812.1 | - | 4 | 804 | ||
AF035287.1 | - | 2 | 94 | |||
AF109126.1 | - | 4 | 779 | |||
AF109127.1 | - | 3 | 431 | |||
AK129908.1 | - | 4 | 359 | |||
AK297184.1 | - | 4 | 703 | |||
AK313441.1 | - | 4 | 670 | |||
AK314248.1 | - | 3 | 323 | |||
AL117411.1 | - | 5 | 857 | |||
BC030773.2 | - | 3 | 322 | |||
BC067828.1 | - | 3 | 482 | |||
BC105979.1 | - | 3 | 403 | 6163693 | ||
BC117462.1 | - | 3 | 321 | 40126013 | ||
BC143880.1 | - | 3 | 321 | 9052396 | ||
BC143881.1 | - | 3 | 321 | 9052397 | ||
CR936779.1 | - | 3 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
