siRNA Details
Gene Symbol: KLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013431.2 | NP_038459.1 | 4 | 675 | ||
GenBank mRNA | AJ001683.1 | - | 4 | 675 | ||
BC017784.1 | - | 4 | 507 | 4619154 | ||
U96845.1 | - | 4 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
