siRNA Details
Gene Symbol: INSRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014215.2 | NP_055030.1 | 3 | 1100 | ||
GenBank mRNA | AK310760.1 | - | 3 | 1101 | ||
J05046.1 | - | 2 | 716 | |||
JQ991924.1 | - | 1 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
